Assay Details
Target Gene Details
Entrez Gene ID: | 10978 |
Gene Name: | cleavage and polyadenylation factor I subunit 1 |
Gene Aliases: |
HEAB, hClp1 |
Location: |
Chr.11:57657744-57661865 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLP1 | NM_001142597.1 | 1 | 71 | NP_001136069.1 |
| NM_006831.2 | 1 | 71 | NP_006822.1 | |
| AK300232.1 | 1 | 54 | ||
| AK313007.1 | 1 | 56 | ||
| BC000446.1 | 1 | 59 | AAH00446.1 | |
| DA922553.1 | 1 | 71 | ||
| U73524.1 | 1 | 657 | AAC50780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
|
| nsv508633 | Chr.11:57652112 - 57792573 on Build GRCh38 | Deletion |
|
| nsv832164 | Chr.11:57640144 - 57811353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map