Assay Details
Target Gene Details
Entrez Gene ID: | 51199 |
Gene Name: | ninein |
Gene Aliases: |
SCKL7 |
Location: |
Chr.14:50719763-50831121 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIN | NM_020921.3 | 31 | 9729 | NP_065972.3 |
| XM_011536817.1 | 31 | 9703 | XP_011535119.1 | |
| XM_011536822.1 | 30 | 7564 | XP_011535124.1 | |
| AB046785.2 | 31 | 7573 | BAB13391.2 | |
| AL359571.1 | 1 | 2159 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564840 | Chr.14:50704161 - 50733474 on Build GRCh38 | Gain |
|
| nsv1040775 | Chr.14:50676760 - 50853094 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map