Assay Details
Target Gene Details
Entrez Gene ID: | 55366 |
Gene Name: | leucine rich repeat containing G protein-coupled receptor 4 |
Gene Aliases: |
BNMD17, GPR48 |
Location: |
Chr.11:27365961-27472787 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGR4 | NM_018490.2 | 18 | 4212 | NP_060960.2 |
| AF257182.1 | 18 | 4171 | AAF68989.1 | |
| BC033039.2 | 17 | 4128 | AAH33039.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv467782 | Chr.11:27171168 - 27474614 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74946148] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map