Assay Details
Target Gene Details
Entrez Gene ID: | 29078 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 4 |
Gene Aliases: |
C6orf66, HRPAP20, HSPC125, My013, bA22L21.1 |
Location: |
Chr.6:96889311-96897891 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF4 | NM_014165.3 | NP_054884.1 | ||
| AF060508.1 | AAG43126.1 | |||
| AF161474.1 | AAF29089.1 | |||
| AK026245.1 | ||||
| AK311850.1 | ||||
| BC039464.1 | AAH39464.1 | |||
| DB253189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv981168 | Chr.6:96889442 - 96891941 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75650763] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map