Assay Details
Target Gene Details
Entrez Gene ID: | 57574 |
Gene Name: | membrane associated ring-CH-type finger 4 |
Gene Aliases: |
MARCH-IV, RNF174 |
Location: |
Chr.2:216257862-216372027 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH4 | NM_020814.2 | 4 | 4299 | NP_065865.1 |
| AB037820.1 | 4 | 4299 | ||
| AL353945.1 | 1 | 921 | ||
| AW051292.1 | 1 | 169 | ||
| BC048793.1 | 4 | 2373 | AAH48793.1 | |
| BC098448.1 | 4 | 2728 | AAH98448.1 | |
| U79281.1 | 1 | 628 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005697 | Chr.2:216231371 - 216429101 on Build GRCh38 | Loss |
|
| nsv1011506 | Chr.2:215969945 - 216310768 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map