Assay Details
Target Gene Details
Entrez Gene ID: | 23032 |
Gene Name: | ubiquitin specific peptidase 33 |
Gene Aliases: |
VDU1 |
Location: |
Chr.1:77695987-77759879 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP33 | NM_015017.4 | NP_055832.3 | ||
| NM_201624.2 | NP_963918.1 | |||
| NM_201626.2 | NP_963920.1 | |||
| XM_005270648.1 | XP_005270705.1 | |||
| XM_005270649.1 | XP_005270706.1 | |||
| XM_011541055.1 | XP_011539357.1 | |||
| XM_011541056.2 | XP_011539358.1 | |||
| XM_017000722.1 | XP_016856211.1 | |||
| XM_017000723.1 | XP_016856212.1 | |||
| XM_017000724.1 | XP_016856213.1 | |||
| XM_017000725.1 | XP_016856214.1 | |||
| XM_017000726.1 | XP_016856215.1 | |||
| XM_017000727.1 | XP_016856216.1 | |||
| XM_017000728.1 | XP_016856217.1 | |||
| XM_017000729.1 | XP_016856218.1 | |||
| AB029020.1 | ||||
| AF383172.1 | AAL78314.1 | |||
| AF383173.1 | AAL78315.1 | |||
| AK022864.1 | BAB14279.1 | |||
| AK023664.1 | ||||
| AK225499.1 | ||||
| AK308402.1 | 12 | 1519 | ||
| BC016663.1 | AAH16663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470719 | Chr.1:77308728 - 77803522 on Build GRCh38 | Gain |
|
| nsv508326 | Chr.1:77718694 - 77768562 on Build GRCh38 | Deletion |
|
| nsv830303 | Chr.1:77625139 - 77843080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map