Assay Details
Target Gene Details
Entrez Gene ID: | 57680 |
Gene Name: | chromodomain helicase DNA binding protein 8 |
Gene Aliases: |
AUTS18, HELSNF1 |
Location: |
Chr.14:21385194-21437298 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD8 | NM_001170629.1 | 26 | 5056 | NP_001164100.1 |
| NM_020920.3 | 27 | 4301 | NP_065971.2 | |
| AB046784.3 | 26 | 4544 | BAB13390.3 | |
| AK056639.1 | 7 | 923 | ||
| AK122910.1 | 27 | 4249 | ||
| AK131077.1 | 12 | 1880 | BAC85127.1 | |
| BC098452.1 | AAH98452.1 | |||
| CR749315.1 | 27 | 4246 | CAH18170.1 |
More Information
Set Membership: |
|

Genomic Map