Assay Details
Target Gene Details
Entrez Gene ID: | 10247 |
Gene Name: | reactive intermediate imine deaminase A homolog |
Gene Aliases: |
HRSP12, P14.5, PSP, UK114 |
Location: |
Chr.8:98102339-98117190 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIDA | NM_005836.2 | NP_005827.1 | ||
| BC010280.1 | AAH10280.1 | |||
| BC012592.1 | AAH12592.1 | |||
| BC093059.1 | AAH93059.1 | |||
| CR456844.1 | CAG33125.1 | |||
| CR541652.1 | CAG46453.1 | |||
| X95384.1 | CAA64670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611806 | Chr.8:98076023 - 98105304 on Build GRCh38 | Gain |
|
| nsv465747 | Chr.8:98093498 - 98127382 on Build GRCh38 | Gain |
|
| nsv831405 | Chr.8:98083011 - 98248706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map