Assay Details
Target Gene Details
Entrez Gene ID: | 254251 |
Gene Name: | ligand dependent nuclear receptor corepressor like |
Gene Aliases: |
MLR1 |
Location: |
Chr.4:17841199-18023287 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCORL | NM_153686.8 | 7 | 1312 | NP_710153.2 |
| NR_136669.1 | 5 | 1138 | ||
| XM_011513821.2 | 9 | 6043 | XP_011512123.1 | |
| XM_011513822.2 | 8 | 6016 | XP_011512124.1 | |
| XM_011513823.2 | 8 | 5889 | XP_011512125.1 | |
| XM_011513826.2 | 6 | 1096 | XP_011512128.1 | |
| XM_017007961.1 | 8 | 7325 | XP_016863450.1 | |
| XM_017007962.1 | 8 | 5967 | XP_016863451.1 | |
| XM_017007963.1 | 8 | 6270 | XP_016863452.1 | |
| XM_017007965.1 | 6 | 2371 | XP_016863454.1 | |
| AK096181.1 | 8 | 1173 | ||
| AK299939.1 | ||||
| BC037322.1 | 7 | 1195 | AAH37322.3 | |
| BE254250.1 | 1 | 304 | ||
| BM721547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819053 | Chr.4:17845457 - 17845767 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map