Assay Details
Target Gene Details
Entrez Gene ID: | 1973 |
Gene Name: | eukaryotic translation initiation factor 4A1 |
Gene Aliases: |
DDX2A, EIF-4A, EIF4A, eIF-4A-I, eIF4A-I |
Location: |
Chr.17:7572706-7579006 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4A1 | NM_001204510.1 | NP_001191439.1 | ||
| NM_001416.3 | NP_001407.1 | |||
| AK130082.1 | ||||
| AK289453.1 | ||||
| AK291971.1 | ||||
| AK296664.1 | ||||
| AK297916.1 | ||||
| AK303222.1 | ||||
| AK309274.1 | ||||
| AK312630.1 | ||||
| BC009585.1 | AAH09585.1 | |||
| BC073752.1 | AAH73752.1 | |||
| BT019880.1 | AAV38683.1 | |||
| BT019881.1 | AAV38684.1 | |||
| D13748.1 | BAA02897.1 |
Target Gene Details
Entrez Gene ID: | 100533955 |
Gene Name: | SENP3-EIF4A1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.17:7561992-7579006 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP3-EIF4A1 | NR_037926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv960401 | Chr.17:7577078 - 7578770 on Build GRCh38 | Duplication |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map