Assay Details
Target Gene Details
Entrez Gene ID: | 55719 |
Gene Name: | SMC5-SMC6 complex localization factor 2 |
Gene Aliases: |
C10orf6, FAM178A |
Location: |
Chr.10:100910230-100965134 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLF2 | NM_001136123.1 | NP_001129595.1 | ||
| NM_018121.3 | NP_060591.3 | |||
| XM_005269965.2 | XP_005270022.1 | |||
| XM_011539944.2 | XP_011538246.1 | |||
| AF460991.1 | AAN84475.1 | |||
| AK001374.1 | BAA91657.1 | |||
| AK057741.1 | 4 | 1148 | BAB71557.1 | |
| AK292565.1 | ||||
| AL832343.1 | ||||
| AL832383.1 | ||||
| AL833126.1 | 6 | 2190 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss |
|
| nsv552051 | Chr.10:100904892 - 100984619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map