Assay Details
Target Gene Details
Entrez Gene ID: | 23074 |
Gene Name: | UHRF1 binding protein 1 like |
Gene Aliases: |
SHIP164 |
Location: |
Chr.12:100026251-100142864 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHRF1BP1L | NM_015054.1 | 21 | 4936 | NP_055869.1 |
| XM_005268738.2 | 22 | 5042 | XP_005268795.1 | |
| XM_005268739.3 | 20 | 4713 | XP_005268796.1 | |
| XM_011538054.2 | 11 | 3411 | XP_011536356.1 | |
| XM_017019047.1 | XP_016874536.1 | |||
| XM_017019048.1 | 21 | 4859 | XP_016874537.1 | |
| XM_017019050.1 | 21 | 4841 | XP_016874539.1 | |
| XM_017019051.1 | 22 | 4987 | XP_016874540.1 | |
| AB014601.2 | 17 | 4377 | BAA31676.2 | |
| AL136821.1 | 14 | 3741 | CAB66755.1 | |
| BC003391.1 | 7 | 1356 | AAH03391.1 | |
| BX647380.1 | 15 | 3907 | CAH56181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2826n54 | Chr.12:99884262 - 100152272 on Build GRCh38 | Loss |
|
| esv3892256 | Chr.12:100007917 - 100225312 on Build GRCh38 | Loss |
|
| nsv523049 | Chr.12:100017297 - 100039015 on Build GRCh38 | Loss |
|
| esv3630541 | Chr.12:99884244 - 100071236 on Build GRCh38 | Gain |
|
| nsv559964 | Chr.12:99872025 - 100538490 on Build GRCh38 | Gain |
|
| nsv559994 | Chr.12:100002546 - 100057456 on Build GRCh38 | Loss |
|

Genomic Map