Assay Details
Target Gene Details
Entrez Gene ID: | 158038 |
Gene Name: | leucine rich repeat and Ig domain containing 2 |
Gene Aliases: |
LERN3, LRRN6C |
Location: |
Chr.9:27937617-29213981 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINGO2 | NM_001258282.1 | 7 | 1161 | NP_001245211.1 |
| NM_152570.2 | 7 | 1241 | NP_689783.1 | |
| XM_011517719.1 | 6 | 2109 | XP_011516021.1 | |
| XM_011517724.2 | 5 | 5128 | XP_011516026.1 | |
| XM_011517728.2 | 2 | 4907 | XP_011516030.1 | |
| XM_017014303.1 | 11 | 5012 | XP_016869792.1 | |
| XM_017014304.1 | 4 | 7532 | XP_016869793.1 | |
| XM_017014305.1 | 5 | 7115 | XP_016869794.1 | |
| XM_017014306.1 | 3 | 4276 | XP_016869795.1 | |
| XM_017014307.1 | 2 | 1749 | XP_016869796.1 | |
| AK056372.1 | 7 | 1241 | BAB71167.1 | |
| AK172834.1 | 1 | 241 | BAD18799.1 | |
| AK290972.1 | 2 | 772 | ||
| AY358114.1 | 2 | 795 | AAQ88481.1 | |
| BC137514.1 | 7 | 1101 | ||
| BC144676.1 | 7 | 1101 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv613950 | Chr.9:27746787 - 28342181 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map