Assay Details
Target Gene Details
Entrez Gene ID: | 5775 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 4 |
Gene Aliases: |
MEG, PTPMEG, PTPMEG1 |
Location: |
Chr.2:119759631-119984899 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN4 | NM_002830.3 | 1 | 444 | NP_002821.1 |
| XM_011511560.2 | 1 | 328 | XP_011509862.1 | |
| XM_011511562.2 | 1 | 328 | XP_011509864.1 | |
| XM_017004596.1 | 1 | 328 | XP_016860085.1 | |
| XM_017004598.1 | 1 | 328 | XP_016860087.1 | |
| XM_017004601.1 | 1 | 328 | XP_016860090.1 | |
| XM_017004602.1 | 1 | 328 | XP_016860091.1 | |
| XM_017004604.1 | 1 | 328 | XP_016860093.1 | |
| XM_017004605.1 | 1 | 328 | XP_016860094.1 | |
| XM_017004606.1 | 1 | 328 | XP_016860095.1 | |
| XM_017004607.1 | 1 | 328 | XP_016860096.1 | |
| XM_017004608.1 | 1 | 328 | XP_016860097.1 | |
| AK314836.1 | 1 | 256 | ||
| BC010674.1 | 1 | 100 | AAH10674.1 | |
| M68941.1 | 1 | 444 | AAA36530.1 |
More Information
Set Membership: |
|

Genomic Map