Assay Details
Target Gene Details
Entrez Gene ID: | 2195 |
Gene Name: | FAT atypical cadherin 1 |
Gene Aliases: |
CDHF7, CDHR8, FAT, ME5, hFat1 |
Location: |
Chr.4:186587783-186726915 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAT1 | NM_005245.3 | 27 | 14021 | NP_005236.2 |
| XM_005262834.2 | 28 | 14449 | XP_005262891.1 | |
| XM_005262835.2 | 28 | 14074 | XP_005262892.1 | |
| XM_006714139.2 | 27 | 14413 | XP_006714202.1 | |
| X87241.1 | 27 | 14025 | CAA60685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025244 | Chr.4:186498616 - 187363659 on Build GRCh38 | Gain |
|
| dgv5495n100 | Chr.4:186554708 - 186992197 on Build GRCh38 | Gain |
|
| nsv596417 | Chr.4:186341335 - 186608750 on Build GRCh38 | Gain |
|
| nsv525719 | Chr.4:186490143 - 186599883 on Build GRCh38 | Gain |
|
| nsv1015619 | Chr.4:186521299 - 186673904 on Build GRCh38 | Gain |
|
| nsv1022361 | Chr.4:186346140 - 188438508 on Build GRCh38 | Gain |
|
| dgv5494n100 | Chr.4:186323525 - 186613274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map