Assay Details
Target Gene Details
Entrez Gene ID: | 54823 |
Gene Name: | SWT1, RNA endoribonuclease homolog |
Gene Aliases: |
C1orf26, HsSwt1 |
Location: |
Chr.1:185157023-185291781 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SWT1 | NM_001105518.1 | 2 | 154 | NP_001098988.1 |
| NM_017673.6 | 2 | 205 | NP_060143.4 | |
| XM_005245271.1 | 2 | 191 | XP_005245328.1 | |
| XM_005245273.1 | 2 | 92 | XP_005245330.1 | |
| XM_011509657.1 | 2 | 171 | XP_011507959.1 | |
| XM_017001527.1 | 2 | 92 | XP_016857016.1 | |
| XM_017001528.1 | 2 | 191 | XP_016857017.1 | |
| XM_017001529.1 | 2 | 85 | XP_016857018.1 | |
| XM_017001530.1 | 2 | 191 | XP_016857019.1 | |
| AF288392.1 | 1 | 51 | AAG60612.1 | |
| AK127980.1 | 2 | 92 | ||
| AK291443.1 | 2 | 71 | ||
| AK315802.1 | 2 | 171 | ||
| BC030781.2 | 2 | 207 | AAH30781.1 | |
| DB453411.1 | 2 | 154 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548396 | Chr.1:185068972 - 185182246 on Build GRCh38 | Loss |
|
| nsv521613 | Chr.1:185102699 - 185202737 on Build GRCh38 | Loss |
|
| esv3575349 | Chr.1:185128640 - 185163037 on Build GRCh38 | Gain |
|

Genomic Map