Assay Details
Target Gene Details
Entrez Gene ID: | 64425 |
Gene Name: | RNA polymerase I subunit E |
Gene Aliases: |
PAF53, PRAF1 |
Location: |
Chr.9:37485935-37503697 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1E | NM_001282766.1 | NP_001269695.1 | ||
| NM_022490.2 | NP_071935.1 | |||
| XM_005251547.2 | XP_005251604.1 | |||
| AK023452.1 | BAB14579.1 | |||
| AK024032.1 | BAB14791.1 | |||
| AK091294.1 | BAC03629.1 | |||
| AK225050.1 | ||||
| AK301347.1 | ||||
| AK302345.1 | ||||
| AK303167.1 | ||||
| BC001337.1 | AAH01337.1 | |||
| BC014331.1 | AAH14331.1 | |||
| CR457313.1 | CAG33594.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv982216 | Chr.9:37489354 - 37491832 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map