Assay Details
Target Gene Details
Entrez Gene ID: | 2700 |
Gene Name: | gap junction protein alpha 3 |
Gene Aliases: |
CTRCT14, CX46, CZP3 |
Location: |
Chr.13:20138252-20161327 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA3 | NM_021954.3 | 2 | 2117 | NP_068773.2 |
| XM_011535048.2 | 2 | 2024 | XP_011533350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037299 | Chr.13:20009920 - 20141363 on Build GRCh38 | Gain |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map