Assay Details
Target Gene Details
Entrez Gene ID: | 3705 |
Gene Name: | inositol-tetrakisphosphate 1-kinase |
Gene Aliases: |
ITRPK1 |
Location: |
Chr.14:92936914-93115918 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPK1 | XM_017021262.1 | 9 | 1464 | XP_016876751.1 |
| XM_017021263.1 | 9 | 1190 | XP_016876752.1 | |
| XM_017021264.1 | 8 | 1262 | XP_016876753.1 | |
| AF279372.1 | 10 | 1328 | AAG44835.1 | |
| AK293515.1 | ||||
| BC003622.2 | AAH03622.1 | |||
| BC007428.2 | AAH07428.1 | |||
| BC018192.1 | 11 | 1437 | AAH18192.1 | |
| BC037305.2 | 9 | 1354 | AAH37305.2 | |
| U51336.1 | 11 | 1381 | AAC50483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456401 | Chr.14:92940717 - 92960275 on Build GRCh38 | Loss |
|
| nsv1400 | Chr.14:92928206 - 92973049 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map