Assay Details
Target Gene Details
Entrez Gene ID: | 139322 |
Gene Name: | apolipoprotein O like |
Gene Aliases: |
CXorf33, FAM121A, Mic27, UNQ8193 |
Location: |
Chr.X:85003852-85093317 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOOL | NM_198450.5 | NP_940852.3 | ||
| XM_011530859.2 | XP_011529161.1 | |||
| XM_017029271.1 | XP_016884760.1 | |||
| XM_017029272.1 | XP_016884761.1 | |||
| XM_017029273.1 | XP_016884762.1 | |||
| AK130506.1 | ||||
| AY358193.1 | AAQ88560.1 | |||
| BC107096.1 | AAI07097.1 | |||
| BI038188.1 | ||||
| CR749250.1 | CAH18106.1 |
More Information
Set Membership: |
|

Genomic Map