Assay Details
Target Gene Details
Entrez Gene ID: | 9874 |
Gene Name: | tousled like kinase 1 |
Gene Aliases: |
PKU-beta |
Location: |
Chr.2:170990823-171231314 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLK1 | NM_001136554.1 | 21 | 3935 | NP_001130026.1 |
| NM_001136555.1 | 19 | 3706 | NP_001130027.1 | |
| NM_012290.4 | 21 | 4376 | NP_036422.3 | |
| XM_011512238.2 | 20 | 4227 | XP_011510540.1 | |
| XM_017005421.1 | 22 | 4485 | XP_016860910.1 | |
| XM_017005422.1 | 20 | 4297 | XP_016860911.1 | |
| AB004885.1 | 22 | 4179 | BAA20562.1 | |
| BC032657.1 | 21 | 3937 | AAH32657.1 | |
| D50927.2 | 21 | 4009 |
More Information
Set Membership: |
|

Genomic Map