Assay Details
Target Gene Details
Entrez Gene ID: | 80204 |
Gene Name: | F-box protein 11 |
Gene Aliases: |
FBX11, PRMT9, UBR6, UG063H01, VIT1 |
Location: |
Chr.2:47806920-47905793 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO11 | NM_001190274.1 | 12 | 1618 | NP_001177203.1 |
| NM_025133.4 | 12 | 1407 | NP_079409.3 | |
| XM_005264572.4 | 12 | 1612 | XP_005264629.1 | |
| XM_005264573.4 | 12 | 1565 | XP_005264630.1 | |
| XM_017005015.1 | 12 | 1565 | XP_016860504.1 | |
| XM_017005016.1 | 12 | 1431 | XP_016860505.1 | |
| XM_017005017.1 | 12 | 1431 | XP_016860506.1 | |
| AF351618.1 | 12 | 1420 | AAN76518.1 | |
| AK022735.1 | 8 | 920 | BAB14214.1 | |
| AK097734.1 | 2 | 250 | ||
| AK223592.1 | 12 | 1502 | BAD97312.1 | |
| AK292877.1 | 12 | 1457 | ||
| AK297444.1 | 12 | 1592 | ||
| AK304140.1 | 12 | 1322 | ||
| AK315108.1 | 12 | 1407 | ||
| AY827075.1 | 12 | 1545 | AAV87312.1 | |
| BC043258.1 | 12 | 1509 | AAH43258.2 | |
| BC130445.1 | 12 | 1343 | ||
| BC136480.1 | 12 | 1343 |
More Information
Set Membership: |
|

Genomic Map