Assay Details
Target Gene Details
Entrez Gene ID: | 80853 |
Gene Name: | lysine demethylase 7A |
Gene Aliases: |
JHDM1D |
Location: |
Chr.7:140084746-140176941 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM7A | NM_030647.1 | 20 | 7456 | NP_085150.1 |
| XM_011516587.2 | 14 | 6481 | XP_011514889.1 | |
| AB051505.1 | 8 | 5761 | BAB21809.1 | |
| AK057208.1 | 1 | 441 | ||
| BX641017.1 | 13 | 6330 | CAE46011.1 | |
| BX649115.1 | 15 | 6723 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map