Assay Details
Target Gene Details
Entrez Gene ID: | 150274 |
Gene Name: | HscB mitochondrial iron-sulfur cluster cochaperone |
Gene Aliases: |
DNAJC20, HSC20, JAC1 |
Location: |
Chr.22:28741992-28757518 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSCB | NM_001318314.1 | NP_001305243.1 | ||
| NM_001318315.1 | NP_001305244.1 | |||
| NM_001318316.1 | NP_001305245.1 | |||
| NM_172002.4 | NP_741999.3 | |||
| NR_134560.1 | ||||
| XM_005261359.3 | XP_005261416.1 | |||
| XM_017028619.1 | XP_016884108.1 | |||
| XM_017028620.1 | XP_016884109.1 | |||
| AY191719.1 | AAN85282.1 | |||
| BC065569.1 | AAH65569.1 | |||
| BG696148.1 | ||||
| BQ054405.1 | ||||
| CR456462.1 | CAG30348.1 | |||
| CR979598.1 | ||||
| HY057929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9895 | Chr.22:28633973 - 28788289 on Build GRCh38 | Gain+Loss |
|
| nsv1064079 | Chr.22:28669540 - 29060071 on Build GRCh38 | Gain |
|
| nsv834169 | Chr.22:28699458 - 28907941 on Build GRCh38 | Loss |
|
| esv3647518 | Chr.22:28747820 - 28768079 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map