Assay Details
Target Gene Details
Entrez Gene ID: | 22920 |
Gene Name: | kinesin associated protein 3 |
Gene Aliases: |
FLA3, KAP-1, KAP-3, KAP3, SMAP, Smg-GDS, dJ190I16.1 |
Location: |
Chr.1:169921329-170074741 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIFAP3 | NM_001204514.1 | NP_001191443.1 | ||
| NM_001204516.1 | NP_001191445.1 | |||
| NM_001204517.1 | NP_001191446.1 | |||
| NM_014970.3 | NP_055785.2 | |||
| XM_005244970.1 | XP_005245027.1 | |||
| XM_011509307.2 | XP_011507609.2 | |||
| AK026471.1 | ||||
| AK122897.1 | ||||
| AK297368.1 | ||||
| AK301912.1 | ||||
| AK303052.1 | ||||
| AK315586.1 | ||||
| BC028679.2 | AAH28679.1 | |||
| U59919.1 | AAC50788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv460n100 | Chr.1:169921401 - 169960441 on Build GRCh38 | Loss |
|
| nsv1010370 | Chr.1:169948309 - 170011810 on Build GRCh38 | Loss |
|
| dgv463n100 | Chr.1:169944456 - 169977295 on Build GRCh38 | Loss |
|
| dgv462n100 | Chr.1:169923520 - 170000303 on Build GRCh38 | Loss |
|
| dgv461n100 | Chr.1:169923143 - 169981200 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map