Assay Details
Target Gene Details
Entrez Gene ID: | 7702 |
Gene Name: | zinc finger protein 143 |
Gene Aliases: |
SBF, STAF, pHZ-1 |
Location: |
Chr.11:9460965-9528524 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF143 | NM_001282656.1 | NP_001269585.1 | ||
| NM_001282657.1 | NP_001269586.1 | |||
| NM_003442.5 | NP_003433.3 | |||
| XM_005253121.1 | XP_005253178.1 | |||
| XM_005253122.1 | XP_005253179.1 | |||
| XM_005253127.1 | XP_005253184.1 | |||
| XM_011520349.2 | XP_011518651.1 | |||
| XM_017018252.1 | XP_016873741.1 | |||
| XM_017018253.1 | XP_016873742.1 | |||
| XM_017018254.1 | XP_016873743.1 | |||
| XM_017018255.1 | XP_016873744.1 | |||
| XM_017018256.1 | XP_016873745.1 | |||
| AF071771.1 | AAC96102.1 | |||
| AK291133.1 | ||||
| AK297214.1 | ||||
| AK313330.1 | ||||
| BC020219.1 | AAH20219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7670 | Chr.11:9479348 - 9493203 on Build GRCh38 | Deletion |
|
| nsv1055078 | Chr.11:9231310 - 9517330 on Build GRCh38 | Gain |
|
| nsv1039537 | Chr.11:9433840 - 9514423 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map