Assay Details
Target Gene Details
Entrez Gene ID: | 292 |
Gene Name: | solute carrier family 25 member 5 |
Gene Aliases: |
2F1, AAC2, ANT2, T2, T3 |
Location: |
Chr.X:119468400-119471396 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A5 | NM_001152.4 | NP_001143.2 | ||
| AK315292.1 | ||||
| BC056160.1 | AAH56160.1 | |||
| BC068199.1 | AAH68199.1 | |||
| J02683.1 | AAA35579.1 | |||
| J03591.1 | AAA36749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740427 | Chr.X:119470076 - 119470389 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map