Assay Details
Target Gene Details
Entrez Gene ID: | 345193 |
Gene Name: | leucine rich repeat, Ig-like and transmembrane domains 3 |
Gene Aliases: |
CSNB1F, FIGLER4 |
Location: |
Chr.4:109848182-109872315 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIT3 | NM_198506.4 | 2 | 367 | NP_940908.3 |
| XM_017008168.1 | 2 | 369 | XP_016863657.1 | |
| AK126648.1 | 1 | 407 | BAC86628.1 | |
| BC104037.2 | AAI04038.1 | |||
| DN994055.1 | 1 | 235 | ||
| DT932743.1 | 1 | 204 |
More Information
Set Membership: |
|

Genomic Map