Assay Details
Target Gene Details
Entrez Gene ID: | 7039 |
Gene Name: | transforming growth factor alpha |
Gene Aliases: |
TFGA |
Location: |
Chr.2:70447280-70554015 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFA | NM_001099691.2 | 6 | 2167 | NP_001093161.1 |
| NM_001308158.1 | 6 | 2337 | NP_001295087.1 | |
| NM_001308159.1 | 6 | 2334 | NP_001295088.1 | |
| NM_003236.3 | 6 | 2170 | NP_003227.1 | |
| AF149096.1 | 6 | 682 | AAF05089.1 | |
| AF149097.1 | 6 | 679 | AAF05090.1 | |
| AF149098.1 | 7 | 711 | AAF05091.1 | |
| BM453873.1 | 1 | 275 | ||
| DA373586.1 | 1 | 228 | ||
| X70340.1 | 6 | 1953 | CAA49806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834255 | Chr.2:70412074 - 70596669 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map