Assay Details
Target Gene Details
Entrez Gene ID: | 280664 |
Gene Name: | WAP four-disulfide core domain 10B |
Gene Aliases: |
WAP12 |
Location: |
Chr.20:45684651-45708899 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC10B | NM_172006.2 | 4 | 569 | NP_742003.1 |
| NM_172131.2 | 3 | 414 | NP_742143.1 | |
| XM_017027824.1 | 5 | 641 | XP_016883313.1 | |
| XM_017027825.1 | 5 | 894 | XP_016883314.1 | |
| AF454506.1 | 4 | 569 | AAN70989.1 | |
| AF454507.1 | 3 | 414 | AAN70990.1 | |
| BC121071.1 | ||||
| BC121072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055601 | Chr.20:45664548 - 45687696 on Build GRCh38 | Loss |
|
| nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map