Assay Details
Target Gene Details
Entrez Gene ID: | 126637 |
Gene Name: | trichohyalin like 1 |
Gene Aliases: |
S100A17, THHL1 |
Location: |
Chr.1:152084144-152089064 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCHHL1 | NM_001008536.1 | NP_001008536.1 | ||
| AY456639.1 | AAS13485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
| nsv547853 | Chr.1:152036195 - 152264284 on Build GRCh38 | Gain |
|

Genomic Map