Assay Details
Target Gene Details
Entrez Gene ID: | 55173 |
Gene Name: | mitochondrial ribosomal protein S10 |
Gene Aliases: |
MRP-S10, PNAS-122 |
Location: |
Chr.6:42206801-42217907 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS10 | NM_018141.3 | NP_060611.2 | ||
| XM_005249206.2 | XP_005249263.1 | |||
| XM_011514724.2 | XP_011513026.1 | |||
| XM_011514725.2 | XP_011513027.1 | |||
| XM_017010996.1 | XP_016866485.1 | |||
| AF113220.1 | AAG39291.1 | |||
| AF277180.1 | AAK07539.1 | |||
| AK001429.1 | BAA91686.1 | |||
| AK098446.1 | ||||
| AK298219.1 | ||||
| AK316599.1 | ||||
| BC012560.1 | AAH12560.1 | |||
| BG776014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830649 | Chr.6:42047596 - 42231697 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map