Assay Details
Target Gene Details
Entrez Gene ID: | 1230 |
Gene Name: | C-C motif chemokine receptor 1 |
Gene Aliases: |
CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1 |
Location: |
Chr.3:46201709-46208341 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR1 | NM_001295.2 | 2 | 2264 | NP_001286.1 |
| AI089760.1 | 1 | 427 | ||
| CK903574.1 | 1 | 118 | ||
| D10925.1 | BAA01723.1 |
More Information
Set Membership: |
|

Genomic Map