Assay Details
Target Gene Details
Entrez Gene ID: | 9134 |
Gene Name: | cyclin E2 |
Gene Aliases: |
CYCE2 |
Location: |
Chr.8:94880224-94896671 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNE2 | NM_057749.2 | 12 | 2254 | NP_477097.1 |
| XM_017013957.1 | 11 | 2579 | XP_016869446.1 | |
| XM_017013958.1 | 12 | 2396 | XP_016869447.1 | |
| XM_017013959.1 | 12 | 2498 | XP_016869448.1 | |
| AF091433.1 | 11 | 2187 | AAC80528.1 |
Target Gene Details
Entrez Gene ID: | 55656 |
Gene Name: | integrator complex subunit 8 |
Gene Aliases: |
C8orf52, INT8 |
Location: |
Chr.8:94823290-94894664 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS8 | XM_017013616.1 | XP_016869105.1 | ||
| XM_017013618.1 | 24 | 3146 | XP_016869107.1 | |
| XM_017013619.1 | 27 | 3552 | XP_016869108.1 | |
| CA867295.1 | 1 | 68 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6314 | Chr.8:94859706 - 94893892 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map