Assay Details
Target Gene Details
Entrez Gene ID: | 8731 |
Gene Name: | RNA guanine-7 methyltransferase |
Gene Aliases: |
CMT1, CMT1c, MET, Met, RG7MT1, cm1p, hCMT1, hMet |
Location: |
Chr.18:13726647-13764556 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNMT | NM_001308263.1 | 8 | 1296 | NP_001295192.1 |
| NM_003799.2 | 8 | 1296 | NP_003790.1 | |
| XM_005258162.1 | 7 | 1167 | XP_005258219.1 | |
| XM_011525749.1 | 8 | 1309 | XP_011524051.1 | |
| XM_011525750.2 | 8 | 1324 | XP_011524052.1 | |
| XM_011525751.1 | 7 | 1167 | XP_011524053.1 | |
| XM_011525753.1 | 7 | 850 | XP_011524055.1 | |
| XM_017026061.1 | 7 | 1167 | XP_016881550.1 | |
| AB007858.1 | 8 | 1251 | BAA23694.1 | |
| AB020966.1 | 7 | 1130 | BAA82447.1 | |
| AB022604.1 | 7 | 1222 | BAA74464.1 | |
| AB022605.1 | 7 | 1222 | BAA74463.1 | |
| AF067791.1 | 8 | 1251 | AAC63269.1 | |
| AK292561.1 | 8 | 1253 | ||
| AK296370.1 | 6 | 702 | ||
| AK303842.1 | ||||
| BC017816.1 | 7 | 1146 | ||
| BC036798.1 | 6 | 1112 | AAH36798.1 | |
| BX647977.1 | 7 | 1147 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576529 | Chr.18:13606185 - 14150725 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map