Assay Details
Target Gene Details
Entrez Gene ID: | 4599 |
Gene Name: | MX dynamin like GTPase 1 |
Gene Aliases: |
IFI-78K, IFI78, MX, MxA |
Location: |
Chr.21:41420329-41459214 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MX1 | NM_001144925.2 | 10 | 1484 | NP_001138397.1 |
| NM_001178046.2 | 6 | 868 | NP_001171517.1 | |
| NM_001282920.1 | 4 | 523 | NP_001269849.1 | |
| NM_002462.4 | 8 | 1043 | NP_002453.2 | |
| XM_005260978.4 | 8 | 959 | XP_005261035.1 | |
| XM_005260979.2 | 8 | 1152 | XP_005261036.1 | |
| XM_005260980.2 | 8 | 924 | XP_005261037.1 | |
| XM_005260981.2 | 7 | 804 | XP_005261038.1 | |
| XM_005260982.2 | 7 | 789 | XP_005261039.1 | |
| XM_011529568.2 | 12 | 1493 | XP_011527870.1 | |
| XM_011529570.2 | 4 | 523 | XP_011527872.1 | |
| XM_017028349.1 | 10 | 1503 | XP_016883838.1 | |
| XM_017028350.1 | 8 | 1171 | XP_016883839.1 | |
| XM_017028351.1 | 8 | 986 | XP_016883840.1 | |
| XM_017028352.1 | 8 | 959 | XP_016883841.1 | |
| AK093008.1 | 4 | 791 | BAC04017.1 | |
| AK096355.1 | 10 | 1527 | ||
| AK225885.1 | 8 | 902 | ||
| AK315465.1 | 6 | 725 | ||
| BC014222.2 | 3 | 252 | AAH14222.2 | |
| BC032602.1 | 7 | 793 | AAH32602.1 | |
| M30817.1 | 6 | 712 | AAA36337.1 | |
| M33882.1 | 8 | 847 | AAA36458.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834102 | Chr.21:41286267 - 41494006 on Build GRCh38 | Loss |
|
| esv3893413 | Chr.21:41410704 - 41454624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map