Assay Details
Target Gene Details
Entrez Gene ID: | 9276 |
Gene Name: | coatomer protein complex subunit beta 2 |
Gene Aliases: |
beta'-COP |
Location: |
Chr.3:139355595-139389680 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPB2 | NM_004766.2 | 14 | 1738 | NP_004757.1 |
| NR_023350.1 | 14 | 1817 | ||
| XM_017007512.1 | 15 | 1923 | XP_016863001.1 | |
| XM_017007513.1 | 13 | 1484 | XP_016863002.1 | |
| AK302943.1 | 14 | 1651 | ||
| BC000326.2 | 14 | 1684 | AAH00326.1 | |
| X70476.1 | 14 | 1676 | CAA49900.1 |
More Information
Set Membership: |
|

Genomic Map