Assay Details
Target Gene Details
Entrez Gene ID: | 105373764 |
Gene Name: | uncharacterized LOC105373764 |
Gene Aliases: |
- |
Location: |
Chr.2:177673322-177723289 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373764 | NR_136171.1 | |||
| BX094187.1 |
Target Gene Details
Entrez Gene ID: | 50940 |
Gene Name: | phosphodiesterase 11A |
Gene Aliases: |
PPNAD2 |
Location: |
Chr.2:177623249-178108339 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE11A | NM_001077196.1 | NP_001070664.1 | ||
| NM_001077197.1 | NP_001070665.1 | |||
| NM_001077358.1 | NP_001070826.1 | |||
| NM_016953.3 | NP_058649.3 | |||
| AB036704.1 | BAB16371.1 | |||
| AB038041.1 | BAB16372.1 | |||
| AF281865.1 | AAG32023.1 | |||
| AJ251509.1 | CAB82573.1 | |||
| AJ278682.1 | CAC15567.1 | |||
| AK074273.1 | ||||
| BC112393.1 | ||||
| BC114431.1 | ||||
| HM771389.1 | ||||
| HM771390.1 | ||||
| HM771391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4109n100 | Chr.2:177671194 - 177721880 on Build GRCh38 | Gain |
|
| esv3593466 | Chr.2:177618127 - 177839958 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map