Assay Details
Target Gene Details
Entrez Gene ID: | 64960 |
Gene Name: | mitochondrial ribosomal protein S15 |
Gene Aliases: |
DC37, MPR-S15, RPMS15, S15mt |
Location: |
Chr.1:36455761-36464439 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS15 | NM_031280.3 | NP_112570.2 | ||
| AB049946.1 | BAB40999.1 | |||
| AB209905.1 | 5 | 461 | BAD93142.1 | |
| AF265439.1 | AAG44697.1 | |||
| AK021626.1 | ||||
| AK302633.1 | 5 | 506 | ||
| AK315441.1 | ||||
| BC031336.1 | AAH31336.1 | |||
| BF310592.1 | ||||
| BM048518.1 | ||||
| BU927793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509202 | Chr.1:36433286 - 36490168 on Build GRCh38 | Insertion |
|
| nsv461073 | Chr.1:36459075 - 36495025 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map