Assay Details
Target Gene Details
Entrez Gene ID: | 80852 |
Gene Name: | glutamate receptor interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.3:14489111-14602996 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIP2 | NM_001080423.3 | NP_001073892.3 | ||
| AB051506.2 |
Target Gene Details
Entrez Gene ID: | 6533 |
Gene Name: | solute carrier family 6 member 6 |
Gene Aliases: |
TAUT |
Location: |
Chr.3:14402576-14489349 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A6 | NM_001134367.3 | 15 | 6312 | NP_001127839.2 |
| NM_003043.5 | 15 | 6316 | NP_003034.2 | |
| NR_103507.2 | 14 | 6191 | ||
| XM_006713307.2 | 14 | 6120 | XP_006713370.1 | |
| XM_011534030.2 | 14 | 6073 | XP_011532332.1 | |
| XM_011534031.2 | 16 | 6444 | XP_011532333.1 | |
| AB209172.1 | 14 | 6117 | BAD92409.1 | |
| CA448606.1 | 1 | 231 | ||
| CD366732.1 | 1 | 231 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2724945 | Chr.3:13931758 - 14820330 on Build GRCh38 | Deletion |
|
| nsv3712 | Chr.3:14467853 - 14501724 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map