Assay Details
Target Gene Details
Entrez Gene ID: | 9064 |
Gene Name: | mitogen-activated protein kinase kinase kinase 6 |
Gene Aliases: |
ASK2, MAPKKK6, MEKK6 |
Location: |
Chr.1:27355179-27368037 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K6 | NM_001297609.1 | NP_001284538.1 | ||
| NM_004672.4 | NP_004663.3 | |||
| XM_017002771.1 | XP_016858260.1 | |||
| AB167411.1 | BAD12485.1 | |||
| AB208805.1 | BAD92042.1 | |||
| AF100318.1 | AAD05304.1 | |||
| AI658745.1 | ||||
| BC015914.1 | AAH15914.1 | |||
| BC129950.1 | ||||
| BC129951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76570472] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map