Assay Details
Target Gene Details
Entrez Gene ID: | 7424 |
Gene Name: | vascular endothelial growth factor C |
Gene Aliases: |
Flt4-L, LMPH1D, VRP |
Location: |
Chr.4:176683534-176792745 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VEGFC | NM_005429.4 | 7 | 1828 | NP_005420.1 |
| AW294939.1 | 1 | 276 | ||
| BC035212.1 | 7 | 1659 | AAH35212.1 | |
| BC063685.1 | 7 | 1810 | AAH63685.1 | |
| U43142.1 | 7 | 1765 | AAA85214.1 | |
| U58111.1 | 7 | 1717 | AAB02909.1 | |
| X94216.1 | 7 | 1745 | CAA63907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion |
|
| nsv4613 | Chr.4:176668312 - 176699170 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map