Assay Details
Target Gene Details
Entrez Gene ID: | 64333 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
10C, RGL1 |
Location: |
Chr.12:57472255-57488814 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP9 | NM_001080156.2 | 16 | 2297 | NP_001073625.1 |
| NM_001080157.1 | 16 | 2546 | NP_001073626.1 | |
| NM_001319850.1 | 21 | 2880 | NP_001306779.1 | |
| NM_001319851.1 | 15 | 2573 | NP_001306780.1 | |
| NM_001319852.1 | 16 | 2354 | NP_001306781.1 | |
| NM_032496.3 | 18 | 2578 | NP_115885.2 | |
| XM_005269083.2 | XP_005269140.1 | |||
| XM_005269084.2 | XP_005269141.1 | |||
| XM_005269085.2 | XP_005269142.1 | |||
| XM_011538656.2 | XP_011536958.1 | |||
| XM_011538657.2 | XP_011536959.1 | |||
| XM_011538658.2 | XP_011536960.1 | |||
| XM_011538659.2 | XP_011536961.1 | |||
| XM_017019800.1 | XP_016875289.1 | |||
| XM_017019801.1 | XP_016875290.1 | |||
| XM_017019802.1 | XP_016875291.1 | |||
| XM_017019803.1 | XP_016875292.1 | |||
| XM_017019804.1 | XP_016875293.1 | |||
| AB030239.1 | BAB83128.1 | |||
| AB051853.1 | BAB56159.1 | |||
| AF161339.1 | AAF28899.1 | |||
| AK057573.1 | ||||
| AK092763.1 | BAC03969.1 | |||
| AK131415.1 | BAD18562.1 | |||
| AK301095.1 | ||||
| AL713803.1 | CAD28552.1 | |||
| BC006107.1 | AAH06107.1 | |||
| BC011820.1 | AAH11820.1 | |||
| CB528354.1 | 1 | 59 |
Target Gene Details
Entrez Gene ID: | 2735 |
Gene Name: | GLI family zinc finger 1 |
Gene Aliases: |
GLI |
Location: |
Chr.12:57460135-57472268 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLI1 | NM_001160045.1 | NP_001153517.1 | ||
| NM_001167609.1 | NP_001161081.1 | |||
| NM_005269.2 | NP_005260.1 | |||
| XM_011538190.1 | XP_011536492.1 | |||
| BC013000.2 | AAH13000.1 | |||
| X07384.1 | CAA30297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759901 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| dgv2651n54 | Chr.12:57450266 - 57552694 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv559035 | Chr.12:57467189 - 57476865 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map