Assay Details
Target Gene Details
Entrez Gene ID: | 10941 |
Gene Name: | UDP glucuronosyltransferase family 2 member A1 complex locus |
Gene Aliases: |
UDPGT2A1 |
Location: |
Chr.4:69588417-69653249 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT2A1 | NM_001252274.2 | 7 | 2807 | NP_001239203.2 |
| NM_001252275.2 | 7 | 2309 | NP_001239204.2 | |
| NM_001301239.1 | 5 | 2115 | NP_001288168.1 | |
| NM_006798.4 | 7 | 2309 | NP_006789.3 | |
| BE463677.1 | 1 | 348 |
Target Gene Details
Entrez Gene ID: | 574537 |
Gene Name: | UDP glucuronosyltransferase family 2 member A2 |
Gene Aliases: |
- |
Location: |
Chr.4:69588417-69639642 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT2A2 | NM_001105677.2 | 6 | 2222 | NP_001099147.2 |
| NM_001301233.1 | 5 | 2090 | NP_001288162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4377 | Chr.4:69578750 - 69638377 on Build GRCh38 | Deletion |
|
| nsv998374 | Chr.4:69581213 - 69624361 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map