Assay Details
Target Gene Details
Entrez Gene ID: | 26133 |
Gene Name: | transient receptor potential cation channel subfamily C member 4 associated protein |
Gene Aliases: |
C20orf188, PPP1R158, TRRP4AP, TRUSS |
Location: |
Chr.20:35002404-35092837 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC4AP | NM_015638.2 | NP_056453.1 | ||
| NM_199368.1 | NP_955400.1 | |||
| XM_011528772.2 | XP_011527074.1 | |||
| XM_011528774.1 | XP_011527076.1 | |||
| XM_017027799.1 | XP_016883288.1 | |||
| XM_017027800.1 | XP_016883289.1 | |||
| XM_017027801.1 | XP_016883290.1 | |||
| AF055022.1 | ||||
| AK074106.1 | BAB84932.1 | |||
| AK298958.1 | ||||
| AK301368.1 | ||||
| AK303476.1 | ||||
| AK315582.1 | ||||
| AL117480.1 | CAB55953.1 | |||
| BC013144.1 | AAH13144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
| nsv833962 | Chr.20:34927688 - 35113250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map