Assay Details
Target Gene Details
Entrez Gene ID: | 124817 |
Gene Name: | cyclin N-terminal domain containing 1 |
Gene Aliases: |
CNTD |
Location: |
Chr.17:42798667-42811587 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTD1 | NM_173478.2 | 7 | 1600 | NP_775749.2 |
| XM_005257043.4 | 7 | 1363 | XP_005257100.1 | |
| XM_005257044.4 | 7 | 1381 | XP_005257101.1 | |
| XM_005257045.4 | 6 | 1523 | XP_005257102.1 | |
| XM_006721690.2 | 7 | 1628 | XP_006721753.1 | |
| XM_011524310.2 | 7 | 1547 | XP_011522612.1 | |
| XM_011524311.2 | 6 | 1205 | XP_011522613.1 | |
| XM_017024171.1 | 6 | 1551 | XP_016879660.1 | |
| AK097456.1 | 7 | 1516 | BAC05060.1 | |
| AK302094.1 | ||||
| AL833052.1 | 5 | 980 | CAH56287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map