Assay Details
Target Gene Details
Entrez Gene ID: | 8284 |
Gene Name: | lysine demethylase 5D |
Gene Aliases: |
HY, HYA, JARID1D, SMCY |
Location: |
Chr.Y:19692495-19745347 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM5D | NM_001146705.1 | NP_001140177.1 | ||
| NM_001146706.1 | NP_001140178.1 | |||
| NM_004653.4 | NP_004644.2 | |||
| XM_005262560.2 | XP_005262617.1 | |||
| XM_005262561.2 | XP_005262618.1 | |||
| XM_011531468.2 | XP_011529770.1 | |||
| AK095923.1 | ||||
| AK127269.1 | ||||
| BC132721.1 | ||||
| BC144102.1 | ||||
| BC146767.1 | ||||
| BX648643.1 | ||||
| D87072.1 | ||||
| U52191.1 | AAC50806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10025 | Chr.Y:18865177 - 20859250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map