Assay Details
Target Gene Details
Entrez Gene ID: | 8661 |
Gene Name: | eukaryotic translation initiation factor 3 subunit A |
Gene Aliases: |
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185 |
Location: |
Chr.10:119035029-119080822 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3A | NM_003750.2 | 19 | 3057 | NP_003741.1 |
| AK302575.1 | 19 | 3002 | ||
| AK307610.1 | 19 | 3035 | ||
| AK309542.1 | 6 | 787 | ||
| BC114429.1 | 19 | 2965 | AAI14430.1 | |
| D50929.1 | 19 | 3039 | ||
| JX312508.1 | 19 | 3058 | ||
| U58046.1 | 19 | 3009 | AAB41584.1 | |
| U78311.1 | 19 | 3024 | AAB80695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041093 | Chr.10:118689033 - 119198373 on Build GRCh38 | Gain |
|
| nsv825582 | Chr.10:119000959 - 119079402 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map