Assay Details
Target Gene Details
Entrez Gene ID: | 5579 |
Gene Name: | protein kinase C beta |
Gene Aliases: |
PKC-beta, PKCB, PRKCB1, PRKCB2 |
Location: |
Chr.16:23835979-24220611 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCB | NM_002738.6 | 1 | 272 | NP_002729.2 |
| NM_212535.2 | 1 | 272 | NP_997700.1 | |
| BC036472.1 | 1 | 232 | AAH36472.1 | |
| BI549165.1 | 1 | 255 | ||
| DB456157.1 | 1 | 272 | ||
| M13975.1 | 1 | 139 | AAA60095.1 | |
| X06318.1 | 1 | 212 | CAA29634.1 | |
| X07109.1 | 1 | 212 | CAA30130.1 |
More Information
Set Membership: |
|

Genomic Map