Assay Details
Target Gene Details
Entrez Gene ID: | 55607 |
Gene Name: | protein phosphatase 1 regulatory subunit 9A |
Gene Aliases: |
NRB1, NRBI, Neurabin-I |
Location: |
Chr.7:94907202-95296415 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R9A | NM_001166160.1 | NP_001159632.1 | ||
| NM_017650.2 | NP_060120.2 | |||
| XM_011516381.1 | XP_011514683.1 | |||
| XM_011516383.1 | XP_011514685.1 | |||
| XM_011516389.1 | XP_011514691.1 | |||
| XM_017012394.1 | XP_016867883.1 | |||
| XM_017012395.1 | XP_016867884.1 | |||
| XM_017012397.1 | XP_016867886.1 | |||
| XM_017012399.1 | XP_016867888.1 | |||
| XM_017012400.1 | XP_016867889.1 | |||
| XM_017012401.1 | XP_016867890.1 | |||
| XM_017012402.1 | XP_016867891.1 | |||
| XM_017012403.1 | XP_016867892.1 | |||
| XM_017012404.1 | XP_016867893.1 | |||
| XM_017012407.1 | XP_016867896.1 | |||
| XM_017012409.1 | XP_016867898.1 | |||
| XM_017012410.1 | XP_016867899.1 | |||
| AK096514.1 | ||||
| DA497168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422248 | Chr.7:94543833 - 94955318 on Build GRCh38 | Duplication |
|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
| nsv831063 | Chr.7:94742146 - 94914116 on Build GRCh38 | Gain |
|
| dgv11513n54 | Chr.7:94709551 - 94942020 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142221536] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map